Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125573.4 | NP_200987.1 | 4 - 5 | 1615 | 113 | |
GenBank mRNA | AY059880.1 | - | 4 - 5 | 1468 | 113 | |
AY086864.1 | - | 4 - 5 | 1470 | 113 | ||
AY114669.1 | - | 4 - 5 | 1357 | 113 | ||
Z18242.1 | - | 1443 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |