Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125578.3 | NP_568941.1 | 2 - 3 | 930 | 76 | |
GenBank mRNA | AY054180.1 | - | 2 - 3 | 721 | 76 | |
AY066039.1 | - | 2 - 3 | 490 | 76 | ||
KJ138631.1 | - | 2 - 3 | 490 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |