Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126003.1 | NP_001119475.1 | 1 - 2 | 402 | 85 | |
NM_125586.4 | NP_568945.2 | 1 - 2 | 1540 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |