Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125600.3 | NP_201013.1 | 1 - 2 | 630 | 64 | |
GenBank mRNA | AY050323.1 | - | 1 - 2 | 522 | 64 | |
AY143917.1 | - | 1 - 2 | 395 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |