Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125605.1 | NP_201018.1 | 1 - 2 | 143 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |