Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345543.1 | NP_001318863.1 | 5 - 6 | 1006 | 138 | |
NM_125619.2 | NP_201031.1 | 4 - 5 | 1029 | 138 | ||
GenBank mRNA | DQ056733.1 | - | 4 - 5 | 899 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |