Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125647.4 | NP_568955.1 | 2 - 3 | 323 | 70 | |
GenBank mRNA | AY065072.1 | - | 2 - 3 | 220 | 70 | |
BT000746.1 | - | 2 - 3 | 253 | 70 | ||
BT010391.1 | - | 2 - 3 | 170 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |