Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333383.1 | NP_001321632.1 | 3 - 4 | 517 | 94 | |
NM_103833.5 | NP_175368.2 | 3 - 4 | 554 | 94 | ||
GenBank mRNA | AF503752.1 | - | 3 - 4 | 308 | 94 | |
AY065424.1 | - | 3 - 4 | 452 | 94 | ||
AY094420.1 | - | 3 - 4 | 389 | 94 | ||
BT001970.1 | - | 3 - 4 | 308 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |