Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333383.1 | NP_001321632.1 | 4 - 5 | 574 | 78 | |
NM_103833.5 | NP_175368.2 | 4 - 5 | 611 | 78 | ||
GenBank mRNA | AY065424.1 | - | 4 - 5 | 509 | 78 | |
AY094420.1 | - | 4 - 5 | 446 | 78 | ||
BT001970.1 | - | 4 - 5 | 365 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |