Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198253.1 | NP_001185182.1 | 17 - 18 | 1999 | 86 | |
NM_103855.5 | NP_175390.2 | 17 - 18 | 1968 | 86 | ||
GenBank mRNA | AY062728.1 | - | 17 - 18 | 1924 | 86 | |
AY072306.1 | - | 17 - 18 | 1929 | 86 | ||
AY093342.1 | - | 17 - 18 | 1843 | 86 | ||
BT008393.1 | - | 17 - 18 | 1843 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |