Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123987.1 | NP_001117459.1 | 24 - 25 | 3646 | 65 | |
NM_103891.4 | NP_175425.2 | 24 - 25 | 3646 | 65 | ||
GenBank mRNA | AF178967.1 | - | 24 - 25 | 3501 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |