Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333424.1 | NP_001323380.1 | 4 - 5 | 555 | 78 | |
NM_001333425.1 | NP_001323379.1 | 3 - 4 | 647 | 78 | ||
NM_103902.4 | NP_564562.1 | 4 - 5 | 565 | 78 | ||
GenBank mRNA | BT004060.1 | - | 4 - 5 | 492 | 78 | |
BT005145.1 | - | 4 - 5 | 476 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |