Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333425.1 | NP_001323379.1 | 4 - 5 | 744 | 95 | |
NM_103902.4 | NP_564562.1 | 5 - 6 | 662 | 95 | ||
GenBank mRNA | BT004060.1 | - | 5 - 6 | 589 | 95 | |
BT005145.1 | - | 5 - 6 | 573 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |