Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALATS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198257.1 | NP_001185186.2 | 12 - 13 | 1804 | 129 | |
NM_103905.4 | NP_175439.2 | 12 - 13 | 1894 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |