Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333427.1 | NP_001319189.1 | 14 - 15 | 1487 | 79 | |
NM_001333428.1 | NP_001321239.1 | 14 - 15 | 1420 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |