Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333427.1 | NP_001319189.1 | 17 - 18 | 2263 | 87 | |
NM_001333428.1 | NP_001321239.1 | 17 - 18 | 2196 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |