Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198259.1 | NP_001185188.1 | 16 - 17 | 1701 | 92 | |
NM_001333440.1 | NP_001322424.1 | 15 - 16 | 1931 | 92 | ||
NM_001333441.1 | NP_001322422.1 | 16 - 17 | 1718 | 92 | ||
NM_001333442.1 | NP_001322423.1 | 16 - 17 | 1686 | 92 | ||
NM_103933.6 | NP_564573.3 | 16 - 17 | 1827 | 92 | ||
GenBank mRNA | AK228745.1 | - | 16 - 17 | 1682 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |