Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333451.1 | NP_001320604.1 | 5 - 6 | 1621 | 115 | |
NM_103952.4 | NP_175485.1 | 5 - 6 | 1718 | 115 | ||
GenBank mRNA | AK175623.1 | - | 5 - 6 | 1568 | 115 | |
BT012659.1 | - | 5 - 6 | 1268 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |