Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121001.2 | NP_568215.2 | 5 - 6 | 636 | 57 | |
GenBank mRNA | AY063833.1 | - | 5 - 6 | 636 | 57 | |
AY091309.1 | - | 5 - 6 | 577 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |