Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121010.3 | NP_196535.1 | 3 - 4 | 994 | 63 | |
GenBank mRNA | AY053409.1 | - | 3 - 4 | 914 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |