Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121055.4 | NP_196579.1 | 8 - 9 | 1516 | 66 | |
GenBank mRNA | AY065415.1 | - | 8 - 9 | 1446 | 66 | |
AY096554.1 | - | 8 - 9 | 1356 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |