Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121056.3 | NP_196580.1 | 8 - 9 | 2032 | 85 | |
GenBank mRNA | AB003591.1 | - | 8 - 9 | 1891 | 85 | |
AY062546.1 | - | 8 - 9 | 1905 | 85 | ||
AY093335.1 | - | 8 - 9 | 1824 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |