Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKC;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121065.3 | NP_196589.1 | 8 - 9 | 846 | 100 | |
GenBank mRNA | AF375437.1 | - | 8 - 9 | 823 | 100 | |
AY120690.1 | - | 8 - 9 | 732 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |