Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121076.3 | NP_196600.1 | 1 | 1042 | 85 | |
GenBank mRNA | AK176500.1 | - | 1 | 1042 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |