Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD4;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343114.1 | NP_001332308.1 | 3 - 4 | 632 | 104 | |
NM_121082.5 | NP_196606.3 | 3 - 4 | 632 | 104 | ||
GenBank mRNA | BT012602.1 | - | 3 - 4 | 490 | 104 | |
BT014959.1 | - | 3 - 4 | 421 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |