Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085097.1 | NP_001078566.1 | 2 - 3 | 279 | 76 | |
NM_001203348.1 | NP_001190277.1 | 2 - 3 | 286 | 76 | ||
NM_121086.5 | NP_196610.2 | 2 - 3 | 300 | 76 | ||
GenBank mRNA | AJ506746.1 | - | 2 - 3 | 199 | 76 | |
AY047708.1 | - | 2 - 3 | 266 | 76 | ||
AY065090.1 | - | 2 - 3 | 279 | 76 | ||
BT006269.1 | - | 2 - 3 | 183 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |