Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085097.1 | NP_001078566.1 | 5 - 6 | 431 | 99 | |
NM_121086.5 | NP_196610.2 | 5 - 6 | 452 | 99 | ||
GenBank mRNA | AJ506746.1 | - | 5 - 6 | 351 | 99 | |
AY047708.1 | - | 5 - 6 | 418 | 99 | ||
AY065090.1 | - | 5 - 6 | 431 | 99 | ||
BT006269.1 | - | 5 - 6 | 335 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |