Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121110.2 | NP_196633.2 | 5 - 6 | 1462 | 73 | |
GenBank mRNA | DQ167579.1 | - | 5 - 6 | 1462 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |