Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343137.1 | NP_001332186.1 | 3 - 4 | 927 | 70 | |
NM_121117.2 | NP_568239.1 | 2 - 3 | 1232 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |