Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G10910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121129.4 | NP_196652.1 | 4 - 5 | 910 | 83 | |
GenBank mRNA | AY058835.1 | - | 4 - 5 | 912 | 83 | |
BT004524.1 | - | 4 - 5 | 875 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |