Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161232.2 | NP_001154704.2 | 7 - 8 | 969 | 116 | |
NM_001161233.1 | NP_001154705.1 | 7 - 8 | 928 | 116 | ||
NM_001343149.1 | NP_001332043.1 | 7 - 8 | 931 | 116 | ||
NM_001343150.1 | NP_001332042.1 | 7 - 8 | 931 | 116 | ||
NM_001343151.1 | NP_001332040.1 | 7 - 8 | 931 | 116 | ||
NM_001343152.1 | NP_001332041.1 | 7 - 8 | 928 | 116 | ||
NM_001343153.1 | NP_001332045.1 | 7 - 8 | 928 | 116 | ||
NM_001343154.1 | NP_001332044.1 | 7 - 8 | 928 | 116 | ||
NM_121141.5 | NP_196664.4 | 7 - 8 | 969 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |