Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121143.2 | NP_196666.1 | 1 - 2 | 391 | 137 | |
GenBank mRNA | AY519620.1 | - | 1 - 2 | 339 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |