Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121148.3 | NP_196671.2 | 7 - 8 | 684 | 126 | |
GenBank mRNA | AM410051.1 | - | 7 - 8 | 685 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |