Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121149.4 | NP_196672.3 | 7 - 8 | 1767 | 132 | |
GenBank mRNA | KJ138698.1 | - | 7 - 8 | 1530 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |