Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343177.1 | NP_001330176.1 | 5 - 6 | 767 | 98 | |
NM_121168.5 | NP_568248.2 | 5 - 6 | 778 | 98 | ||
GenBank mRNA | BT026128.1 | - | 4 - 5 | 555 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |