Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085101.2 | NP_001078570.1 | 7 - 8 | 1568 | 92 | |
NM_001343183.1 | NP_001332634.1 | 7 - 8 | 1567 | 92 | ||
NM_001343184.1 | NP_001332633.1 | 7 - 8 | 1567 | 92 | ||
NM_121176.4 | NP_196699.1 | 7 - 8 | 1567 | 92 | ||
GenBank mRNA | AK227249.1 | - | 7 - 8 | 1561 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |