Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121218.5 | NP_196741.2 | 2 - 3 | 525 | 75 | |
GenBank mRNA | BT044605.1 | - | 2 - 3 | 321 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |