Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121277.4 | NP_568272.1 | 2 - 3 | 423 | 108 | |
GenBank mRNA | AB195718.1 | - | 2 - 3 | 239 | 108 | |
AK118241.1 | - | 2 - 3 | 328 | 108 | ||
BT005496.1 | - | 2 - 3 | 239 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |