Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202664.1 | NP_001189593.1 | 8 - 9 | 1262 | 80 | |
NM_128025.4 | NP_565576.1 | 7 - 8 | 1193 | 80 | ||
GenBank mRNA | AF360315.1 | - | 7 - 8 | 1170 | 80 | |
AK319102.1 | - | 8 - 9 | 1235 | 80 | ||
AY056366.1 | - | 7 - 8 | 1080 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |