Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128071.3 | NP_565586.1 | 3 - 4 | 902 | 100 | |
GenBank mRNA | AY070722.1 | - | 3 - 4 | 851 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |