Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335976.1 | NP_001324292.1 | 25 - 26 | 3095 | 153 | |
NM_001335977.1 | NP_001324293.1 | 25 - 26 | 3122 | 153 | ||
NM_001335978.1 | NP_001324291.1 | 24 - 25 | 3044 | 153 | ||
NM_128074.4 | NP_565587.1 | 25 - 26 | 3068 | 153 | ||
GenBank mRNA | AF185577.1 | - | 25 - 26 | 3068 | 153 | |
AF185578.1 | - | 24 - 25 | 3048 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |