Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335976.1 | NP_001324292.1 | 29 - 30 | 3778 | 78 | |
NM_001335977.1 | NP_001324293.1 | 29 - 30 | 3805 | 78 | ||
NM_001335978.1 | NP_001324291.1 | 28 - 29 | 3727 | 78 | ||
NM_128074.4 | NP_565587.1 | 29 - 30 | 3751 | 78 | ||
GenBank mRNA | AF185577.1 | - | 29 - 30 | 3751 | 78 | |
AF185578.1 | - | 28 - 29 | 3731 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |