Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335998.1 | NP_001325074.1 | 1 - 2 | 461 | 146 | |
NM_001335999.1 | NP_001325073.1 | 1 - 2 | 448 | 146 | ||
NM_128104.5 | NP_180117.4 | 1 - 2 | 335 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |