Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336002.1 | NP_001318288.1 | 1 - 2 | 230 | 89 | |
GenBank mRNA | KJ138898.1 | - | 1 - 2 | 64 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |