Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128118.5 | NP_180131.3 | 2 - 3 | 955 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |