Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202675.1 | NP_001189604.1 | 2 | 1330 | 96 | |
NM_128150.5 | NP_180161.1 | 2 | 1440 | 96 | ||
GenBank mRNA | AF375403.1 | - | 1 | 1063 | 96 | |
AK117272.1 | - | 1 | 1057 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |