Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 25 - 26 | 2972 | 68 | |
GenBank mRNA | AY035110.1 | - | 25 - 26 | 2281 | 68 | |
FJ708701.1 | - | 25 - 26 | 2205 | 68 | ||
U47029.1 | - | 25 - 26 | 2255 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |