Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336066.1 | NP_001325287.1 | 4 - 5 | 1112 | 100 | |
NM_128198.5 | NP_180209.4 | 5 - 6 | 1019 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |