Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336067.1 | NP_001318292.1 | 5 - 6 | 2144 | 100 | |
NM_001336068.1 | NP_001324180.1 | 5 - 6 | 2144 | 142 | ||
NM_001336069.1 | NP_001324179.1 | 4 - 5 | 1770 | 142 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |