Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336074.1 | NP_001324267.1 | 7 - 8 | 1145 | 67 | |
NM_128211.4 | NP_565625.1 | 8 - 9 | 1027 | 67 | ||
GenBank mRNA | BT026116.1 | - | 8 - 9 | 891 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |